accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1244855-68-2

H3COOSiH3CCH3OH

CAS RN 1244855-68-2

IUPAC Name: ethyl 4-{[2-(hydroxymethyl)phenyl]dimethylsilyl}benzoate
Synonyms: ethyl 4-{[2-(hydroxymethyl)phenyl]dimethylsilyl}benzoate
Molecular Weight (g/mol): 314.46
Molecular Formula: C18H22O3Si
InChi Key: VPQQJIOOSRZZDJ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(C=C1)[Si](C)(C)C1=CC=CC=C1CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results