accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FOFFFOH

CAS RN 1242252-68-1

IUPAC Name: [4-fluoro-2-(trifluoromethoxy)phenyl]methanol
Synonyms: [4-fluoro-2-(trifluoromethoxy)phenyl]methanol
Molecular Weight (g/mol): 210.13
Molecular Formula: C8H6F4O2
InChi Key: QTZPHLXETXULFE-UHFFFAOYSA-N
SMILES: OCC1=C(OC(F)(F)F)C=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results