accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FOFFFNH2

CAS RN 1241828-99-8

IUPAC Name: 1-[4-fluoro-2-(trifluoromethoxy)phenyl]methanamine
Synonyms: 1-[4-fluoro-2-(trifluoromethoxy)phenyl]methanamine
Molecular Weight (g/mol): 209.14
Molecular Formula: C8H7F4NO
InChi Key: YYVIURGLQBEQSP-UHFFFAOYSA-N
SMILES: NCC1=C(OC(F)(F)F)C=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results