accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OONNHBrFFF

CAS RN 1240278-21-0

IUPAC Name: ethyl 4-bromo-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate
Synonyms: ethyl 4-bromo-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate
Molecular Weight (g/mol): 287.04
Molecular Formula: C7H6BrF3N2O2
InChi Key: OVIHBAGNAYIHMQ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=NNC(=C1Br)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)