accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1240257-81-1

SFFFFFClOCl

CAS RN 1240257-81-1

IUPAC Name: 3-chloro-5-(pentafluoro-λ⁶-sulfanyl)benzoyl chloride
Synonyms: 3-chloro-5-(pentafluoro-λ⁶-sulfanyl)benzoyl chloride
Molecular Weight (g/mol): 301.05
Molecular Formula: C7H3Cl2F5OS
InChi Key: GLJCAZZLRUQUSD-UHFFFAOYSA-N
SMILES: FS(F)(F)(F)(F)C1=CC(Cl)=CC(=C1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results