accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

SFFFFFClN

CAS RN 1240257-75-3

IUPAC Name: 2-[3-chloro-5-(pentafluoro-λ⁶-sulfanyl)phenyl]acetonitrile
Synonyms: 2-[3-chloro-5-(pentafluoro-λ⁶-sulfanyl)phenyl]acetonitrile
Molecular Weight (g/mol): 277.64
Molecular Formula: C8H5ClF5NS
InChi Key: WTFFVDFJXLCKCQ-UHFFFAOYSA-N
SMILES: FS(F)(F)(F)(F)C1=CC(Cl)=CC(CC#N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results