accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FSFFFFFNH2

CAS RN 1240257-73-1

IUPAC Name: 1-[3-fluoro-5-(pentafluoro-λ⁶-sulfanyl)phenyl]methanamine
Synonyms: 1-[3-fluoro-5-(pentafluoro-λ⁶-sulfanyl)phenyl]methanamine
Molecular Weight (g/mol): 251.19
Molecular Formula: C7H7F6NS
InChi Key: FKGCFDAJSOIINF-UHFFFAOYSA-N
SMILES: NCC1=CC(F)=CC(=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results