accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

SFFFFFOHOF

CAS RN 1240257-15-1

IUPAC Name: 2-fluoro-5-(pentafluoro-λ⁶-sulfanyl)benzoic acid
Synonyms: 2-fluoro-5-(pentafluoro-λ⁶-sulfanyl)benzoic acid
Molecular Weight (g/mol): 266.16
Molecular Formula: C7H4F6O2S
InChi Key: HQONRARZDMQSRY-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C=CC(=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results