CAS RN 1240257-14-0
IUPAC Name:
1-bromo-3-(bromomethyl)-5-(pentafluoro-λ⁶-sulfanyl)benzene
Synonyms:
1-bromo-3-(bromomethyl)-5-(pentafluoro-λ⁶-sulfanyl)benzene
Molecular Weight (g/mol):
375.98
Molecular Formula:
C7H5Br2F5S
InChi Key:
PSRNYAZILJJJGP-UHFFFAOYSA-N
SMILES:
FS(F)(F)(F)(F)C1=CC(Br)=CC(CBr)=C1
1
–
1
of
1
results
1
3-Bromo-5-(pentafluorothio)benzyl bromide, 97%, Thermo Scientific™
CAS: 1240257-14-0 Molecular Formula: C7H5Br2F5S Molecular Weight (g/mol): 375.977 MDL Number: MFCD16652484 InChI Key: PSRNYAZILJJJGP-UHFFFAOYSA-N Synonym: 3-bromo-5-pentafluorosulfur benzyl bromide,3-bromo-5-pentafluorothio benzyl bromide,1-bromo-3-bromomethyl-5-pentafluoro-??-sulfanyl benzene PubChem CID: 66523602 IUPAC Name: [3-bromo-5-(bromomethyl)phenyl]-pentafluoro-$l^{6}-sulfane SMILES: C1=C(C=C(C=C1S(F)(F)(F)(F)F)Br)CBr