accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1240257-04-8

NSFFFFF

CAS RN 1240257-04-8

IUPAC Name: 2-[4-(pentafluoro-λ⁶-sulfanyl)phenyl]acetonitrile
Synonyms: 2-[4-(pentafluoro-λ⁶-sulfanyl)phenyl]acetonitrile
Molecular Weight (g/mol): 243.2
Molecular Formula: C8H6F5NS
InChi Key: APQCOBQUUKSALE-UHFFFAOYSA-N
SMILES: FS(F)(F)(F)(F)C1=CC=C(CC#N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results