accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FOFFFNH2

CAS RN 123572-66-7

IUPAC Name: 4-fluoro-2-(trifluoromethoxy)aniline
Synonyms: 4-fluoro-2-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 195.12
Molecular Formula: C7H5F4NO
InChi Key: QFDKCJQODJFZID-UHFFFAOYSA-N
SMILES: NC1=C(OC(F)(F)F)C=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results