accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 123548-16-3

H3CNCH3NONONHN

CAS RN 123548-16-3

IUPAC Name: 2-(2-{4-[4-(diethylamino)butyl]piperidin-1-yl}acetyl)-2,4,9-triazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-10-one
Synonyms: 2-(2-{4-[4-(diethylamino)butyl]piperidin-1-yl}acetyl)-2,4,9-triazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-10-one
Molecular Weight (g/mol): 463.63
Molecular Formula: C27H37N5O2
InChi Key: BCUGCHZRMKTPMU-UHFFFAOYSA-N
SMILES: CCN(CC)CCCCC1CCN(CC(=O)N2C3=CC=CC=C3C(=O)NC3=CC=CN=C23)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results