accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NNOHO

CAS RN 123533-41-5

IUPAC Name: 2-phenylimidazo[1,2-a]pyridine-3-carboxylic acid
Synonyms: 2-phenylimidazo[1,2-a]pyridine-3-carboxylic acid
Molecular Weight (g/mol): 238.25
Molecular Formula: C14H10N2O2
InChi Key: WJWJFXUNWPMQSU-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(N=C2C=CC=CN12)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results