accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 123333-64-2

OHClOClSOOO(Z)

CAS RN 123333-64-2

IUPAC Name: 2-[(3-chloro-4-hydroxyphenyl)[(1Z)-3-chloro-4-oxocyclohexa-2,5-dien-1-ylidene]methyl]benzene-1-sulfonate
Synonyms: 2-[(3-chloro-4-hydroxyphenyl)[(1Z)-3-chloro-4-oxocyclohexa-2,5-dien-1-ylidene]methyl]benzenesulfonate
Molecular Weight (g/mol): 422.25
Molecular Formula: C19H11Cl2O5S
InChi Key: QXTPRQZMDKBTAI-UNOMPAQXSA-M
SMILES: OC1=CC=C(C=C1Cl)C(=C1/C=CC(=O)C(Cl)=C1)\C1=CC=CC=C1S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results