accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 122996-47-8

H3CH3CCH3ONOOOHO

CAS RN 122996-47-8

IUPAC Name: 4-(tert-butoxy)-1-{[(9H-fluoren-9-yl)methoxy]carbonyl}pyrrolidine-2-carboxylic acid
Synonyms: 4-(tert-butoxy)-1-[(9H-fluoren-9-ylmethoxy)carbonyl]pyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 409.48
Molecular Formula: C24H27NO5
InChi Key: WPBXBYOKQUEIDW-UHFFFAOYNA-N
SMILES: CC(C)(C)OC1CC(N(C1)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)