accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

RuCH2H2CCH2H2CCH3CH3

CAS RN 12289-94-0

IUPAC Name: λ²-ruthenium(2+) bis(2-methylprop-2-en-1-ide) cycloocta-1,5-diene
Synonyms: λ²-ruthenium(2+) 1,5-cyclooctadiene bis(2-methylprop-2-en-1-ide)
Molecular Weight (g/mol): 319.45
Molecular Formula: C16H26Ru
InChi Key: POYBJJLKGYXKJH-UHFFFAOYSA-N
SMILES: [Ru++].CC([CH2-])=C.CC([CH2-])=C.C1CC=CCCC=C1
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results