accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 122536-73-6

NH3(R)NOO

CAS RN 122536-73-6

IUPAC Name: (3R)-1-[(benzyloxy)carbonyl]pyrrolidin-3-aminium
Synonyms: (3R)-1-[(benzyloxy)carbonyl]pyrrolidin-3-aminium
Molecular Weight (g/mol): 221.28
Molecular Formula: C12H17N2O2
InChi Key: FPXJNSKAXZNWMQ-LLVKDONJSA-O
SMILES: [NH3+][C@@H]1CCN(C1)C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results