accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 122350-52-1

OOOOHNHOO

CAS RN 122350-52-1

IUPAC Name: 5-(benzyloxy)-4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxopentanoic acid
Synonyms: 5-(benzyloxy)-4-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-5-oxopentanoic acid
Molecular Weight (g/mol): 459.5
Molecular Formula: C27H25NO6
InChi Key: FMWLYDDRYGOYMY-UHFFFAOYNA-N
SMILES: OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)