accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12233-56-6

H2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OGeGeGeBiBiBiBi

CAS RN 12233-56-6

IUPAC Name: tetrabismuth trigermanium dodecahydrate
Synonyms: tetrabismuth trigermanium dodecahydrate
Molecular Weight (g/mol): 1269.99
Molecular Formula: Bi4Ge3H24O12
InChi Key: ORCSMBGZHYTXOV-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.O.O.O.O.O.O.[Ge].[Ge].[Ge].[Bi].[Bi].[Bi].[Bi]
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results