CAS RN 12229-13-9
CAS RN 12229-13-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Potassium pentaborate octahydrate, 97%, Thermo Scientific™
CAS: 12229-13-9 MDL Number: MFCD00213879 Synonym: potassium pentaborate octahydrate, 97