accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1222630-32-1

BFFFFPH3N

CAS RN 1222630-32-1

IUPAC Name: (2-azaniumylethyl)diphenylphosphane; tetrafluoroboranuide
Synonyms: (2-ammonioethyl)diphenylphosphane tetrafluoroborate
Molecular Weight (g/mol): 317.07
Molecular Formula: C14H17BF4NP
InChi Key: ACXZLRCRPLYXBQ-UHFFFAOYSA-O
SMILES: F[B-](F)(F)F.[NH3+]CCP(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)