accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CCH3NH3CCH3POCH3OCH3

CAS RN 122194-07-4

IUPAC Name: (dimethoxyphosphanyl)bis(propan-2-yl)amine
Synonyms: (dimethoxyphosphanyl)diisopropylamine
Molecular Weight (g/mol): 193.23
Molecular Formula: C8H20NO2P
InChi Key: KXUMNSXPAYCKPR-UHFFFAOYSA-N
SMILES: COP(OC)N(C(C)C)C(C)C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)