accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 122023-29-4

H3COFFFF

CAS RN 122023-29-4

IUPAC Name: 1-[2-fluoro-4-(trifluoromethyl)phenyl]ethan-1-one
Synonyms: 1-[2-fluoro-4-(trifluoromethyl)phenyl]ethanone
Molecular Weight (g/mol): 206.14
Molecular Formula: C9H6F4O
InChi Key: OLDXVWXLMGJVPH-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results