accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1218790-84-1

IUPAC Name: [2-cyano-4-(trifluoromethyl)phenyl]boronic acid
Synonyms: 2-cyano-4-(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol): 214.94
Molecular Formula: C8H5BF3NO2
InChi Key: DELAZGQTKSRLPA-UHFFFAOYSA-N
SMILES: OB(O)C1=C(C=C(C=C1)C(F)(F)F)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results