accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BOOH3CCH3H3CH3CSN

CAS RN 1218790-45-4

IUPAC Name: 1-{[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methyl}pyrrolidine
Synonyms: 1-{[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methyl}pyrrolidine
Molecular Weight (g/mol): 293.23
Molecular Formula: C15H24BNO2S
InChi Key: VWOUUXTUYSWOCT-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(CN2CCCC2)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results