CAS RN 1217641-89-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Eletriptan N-Oxide, TRC
CAS: 1217641-89-8 Molecular Formula: C22H26N2O3S Molecular Weight (g/mol): 398.52 Synonym: 3-[[(2R)-1-Methyl-2-pyrrolidinyl]methyl]-5-[2-(phenylsulfonyl)ethyl]-1H-indole N Oxide; IUPAC Name: 5-[2-(benzenesulfonyl)ethyl]-3-[[(2R)-1-methyl-1-oxidopyrrolidin-1-ium-2-yl]methyl]-1H-indole SMILES: C[N+]1([O-])CCC[C@@H]1Cc2c[nH]c3ccc(CCS(=O)(=O)c4ccccc4)cc23