CAS RN 1217500-55-4
CAS RN 1217500-55-4
IUPAC Name:
(3-chloro-2-fluoropyridin-4-yl)boronic acid
Synonyms:
3-chloro-2-fluoropyridin-4-ylboronic acid
Molecular Weight (g/mol):
175.35
Molecular Formula:
C5H4BClFNO2
InChi Key:
VYHSSQRGIWMXJE-UHFFFAOYSA-N
SMILES:
OB(O)C1=C(Cl)C(F)=NC=C1
1
–
1
of
1
results
1
3-Chloro-2-fluoropyridine-4-boronic acid, 95%, Thermo Scientific™
CAS: 1217500-55-4 Molecular Formula: C5H4BClFNO2 Molecular Weight (g/mol): 175.35 MDL Number: MFCD09037477 InChI Key: VYHSSQRGIWMXJE-UHFFFAOYSA-N Synonym: 3-chloro-2-fluoropyridine-4-boronic acid,3-chloro-2-fluoropyridin-4-yl boronic acid,acmc-209af7 PubChem CID: 44717688 IUPAC Name: (3-chloro-2-fluoropyridin-4-yl)boronic acid SMILES: B(C1=C(C(=NC=C1)F)Cl)(O)O