accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12137-27-8

ORhO

CAS RN 12137-27-8

IUPAC Name: dioxorhodium
Synonyms: rhodium(IV) oxide
Molecular Weight (g/mol): 134.9
Molecular Formula: O2Rh
InChi Key: KZYDBKYFEURFNC-UHFFFAOYSA-N
SMILES: O=[Rh]=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results