CAS RN 12136-58-2
CAS RN 12136-58-2
IUPAC Name:
dilithium(1+) sulfanide
Synonyms:
lithium sulfide
Molecular Weight (g/mol):
46.95
Molecular Formula:
HLi2S
InChi Key:
ZEBXXUVDYRCDCF-UHFFFAOYSA-M
SMILES:
[Li+].[Li+].[SH-]
1
–
1
of
1
results
1
Promotions Available
Lithium sulfide, 99.9% (metals basis)
Lithium sulfide, 99.9% (metals basis)
CAS: 12136-58-2 Molecular Formula: Li2S Molecular Weight (g/mol): 45.94 MDL Number: MFCD00011085 InChI Key: GLNWILHOFOBOFD-UHFFFAOYSA-N Synonym: lithium sulfide,lithium sulfide li2s,lithium sulphide, anhydrous,li2s,zwdbutfcwlvlcq-uhfffaoysa-n,lithium sulfide trace metals basis PubChem CID: 64734 SMILES: [Li+].[Li+].[S--]