accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12134-75-7

SiH4SiH4Gd

CAS RN 12134-75-7

IUPAC Name: gadolinium(3+) disilane
Synonyms: gadolinium(3+) disilane
Molecular Weight (g/mol): 221.48
Molecular Formula: GdH8Si2
InChi Key: AOLGUBMBPNHJFD-UHFFFAOYSA-N
SMILES: [SiH4].[SiH4].[Gd+3]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results