accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12125-22-3

PdS

CAS RN 12125-22-3

IUPAC Name: sulfanylidenepalladium
Synonyms: sulfanylidenepalladium
Molecular Weight (g/mol): 138.48
Molecular Formula: PdS
InChi Key: NRUVOKMCGYWODZ-UHFFFAOYSA-N
SMILES: S=[Pd]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results