accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12084-29-6

OOOOBaH3CH3CCH3CH3

CAS RN 12084-29-6

IUPAC Name: barium(2+) bis(2,4-dioxopentan-3-ide)
Synonyms: barium acetylacetonate
Molecular Weight (g/mol): 335.55
Molecular Formula: C10H14BaO4
InChi Key: PCCXJFCCJLNBDF-UHFFFAOYSA-N
SMILES: [Ba++].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results