accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12070-06-3

CTa

CAS RN 12070-06-3

IUPAC Name: methane tantalum
Synonyms: tantalum carbide
Molecular Weight (g/mol): 196.99
Molecular Formula: CH4Ta
InChi Key: SLVPOVIZFDXMPC-UHFFFAOYSA-N
SMILES: C.[Ta]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results