accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12069-94-2

CNb

CAS RN 12069-94-2

IUPAC Name: methane niobium
Synonyms: niobium carbide
Molecular Weight (g/mol): 108.95
Molecular Formula: CH4Nb
InChi Key: CLOIETWQBJYMFR-UHFFFAOYSA-N
SMILES: C.[Nb]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results