accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12068-90-5

TeHg

CAS RN 12068-90-5

IUPAC Name: tellanylidenemercury
Synonyms: mercury telluride
Molecular Weight (g/mol): 328.19
Molecular Formula: HgTe
InChi Key: VCEXCCILEWFFBG-UHFFFAOYSA-N
SMILES: [Te]=[Hg]
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)