accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FeSS

CAS RN 12068-85-8

IUPAC Name: λ²-iron(2+) disulfanediide
Synonyms: λ²-iron(2+) disulfide(2-)
Molecular Weight (g/mol): 119.97
Molecular Formula: FeS2
InChi Key: NIFIFKQPDTWWGU-UHFFFAOYSA-N
SMILES: [Fe++].[S-][S-]
Physical Form 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)