accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12068-61-0

NiAsAs

CAS RN 12068-61-0

IUPAC Name: 1λ²,2λ²-diarsenyl nickel
Synonyms: 1λ²,2λ²-diarsenyl nickel
Molecular Weight (g/mol): 208.54
Molecular Formula: As2Ni
InChi Key: LPZQKJKDQZNVJW-UHFFFAOYSA-N
SMILES: [Ni].[As]=[As]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results