accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12068-51-8

AlMgO24

CAS RN 12068-51-8

IUPAC Name: dialuminium(3+) magnesium(2+) tetraoxidandiide
Synonyms: dialuminium(3+) magnesium(2+) tetraoxidandiide
Molecular Weight (g/mol): 142.26
Molecular Formula: Al2MgO4
InChi Key: UAMZXLIURMNTHD-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[Mg++].[Al+3].[Al+3]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results