accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3ONH2FFF

CAS RN 1206593-27-2

IUPAC Name: 1-[4-ethoxy-3-(trifluoromethyl)phenyl]methanamine
Synonyms: 1-[4-ethoxy-3-(trifluoromethyl)phenyl]methanamine
Molecular Weight (g/mol): 219.21
Molecular Formula: C10H12F3NO
InChi Key: GHCHXTNRZYWWPF-UHFFFAOYSA-N
SMILES: CCOC1=C(C=C(CN)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results