accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12065-68-8

OTaOOPbOTaOO

CAS RN 12065-68-8

IUPAC Name: tetraoxo-2,4-dioxa-3-plumba-1,5-ditantalapentane
Synonyms: tetraoxo-2,4-dioxa-3-plumba-1,5-ditantalapentane
Molecular Weight (g/mol): 665.1
Molecular Formula: O6PbTa2
InChi Key: KNSIKTVGKTWOGA-UHFFFAOYSA-N
SMILES: O=[Ta](=O)O[Pb]O[Ta](=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results