accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12064-03-8

GaSb

CAS RN 12064-03-8

IUPAC Name: gallium(3+) antimony
Synonyms: gallium antimonide
Molecular Weight (g/mol): 191.48
Molecular Formula: GaSb
InChi Key: HDDJZDZAJXHQIL-UHFFFAOYSA-N
SMILES: [Ga+3].[Sb]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results