accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12053-39-3

CrTeCrTeTe

CAS RN 12053-39-3

IUPAC Name: bis(tellanylidenechromium) tellurium
Synonyms: bis(tellanylidenechromium) tellurium
Molecular Weight (g/mol): 486.79
Molecular Formula: Cr2Te3
InChi Key: PDJHBRMODJKXLQ-UHFFFAOYSA-N
SMILES: [Te].[Cr]=[Te].[Cr]=[Te]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results