CAS RN 1205-02-3
IUPAC Name:
2-(phenylformamido)propanoic acid
Synonyms:
methylhippuric acid
Molecular Weight (g/mol):
193.2
Molecular Formula:
C10H11NO3
InChi Key:
UAQVHNZEONHPQG-UHFFFAOYNA-N
SMILES:
CC(NC(=O)C1=CC=CC=C1)C(O)=O
1
–
1
of
1
results
1
N-Benzoyl-DL-alanine, 97+%
CAS: 1205-02-3 Molecular Formula: C10H11NO3 Molecular Weight (g/mol): 193.20 MDL Number: MFCD00020393 InChI Key: UAQVHNZEONHPQG-UHFFFAOYNA-N Synonym: n-benzoyl-dl-alanine,benzoyl-dl-alanine,methylhippuric acid,n-benzoylalanine,alanine, n-benzoyl,benzoylalanine,dl-n-benzoylalanine,2-phenylformamido propanoic acid,alpha-methylhippuric acid,n-phenylcarbonyl alanine PubChem CID: 71002 ChEBI: CHEBI:71167 IUPAC Name: 2-benzamidopropanoic acid SMILES: CC(NC(=O)C1=CC=CC=C1)C(O)=O