accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12042-92-1

OAlOOAlOCu

CAS RN 12042-92-1

IUPAC Name: copper(2+) dialumanoylolate
Synonyms: copper(2+) dialumanoylolate
Molecular Weight (g/mol): 181.51
Molecular Formula: Al2CuO4
InChi Key: IPJRADJOSNLGBF-UHFFFAOYSA-N
SMILES: [Cu++].[O-][Al]=O.[O-][Al]=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)