accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12039-90-6

H2SiZrSiH2

CAS RN 12039-90-6

IUPAC Name: (silanylidenezirconio)silane
Synonyms: zirconium disilicide
Molecular Weight (g/mol): 151.43
Molecular Formula: H4Si2Zr
InChi Key: ZSONHIWWHDAZQO-UHFFFAOYSA-N
SMILES: [SiH2]=[Zr]=[SiH2]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results