accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12039-88-2

SiWSi

CAS RN 12039-88-2

IUPAC Name: tungsten disilicide
Synonyms: tungsten disilicide
Molecular Weight (g/mol): 240.01
Molecular Formula: Si2W
InChi Key: WQJQOUPTWCFRMM-UHFFFAOYSA-N
SMILES: [Si]#[W]#[Si]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results