accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12036-02-1

OOsO

CAS RN 12036-02-1

IUPAC Name: dioxoosmium
Synonyms: osmium dioxide
Molecular Weight (g/mol): 222.23
Molecular Formula: O2Os
InChi Key: XSXHWVKGUXMUQE-UHFFFAOYSA-N
SMILES: O=[Os]=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results