accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12035-51-7

NiSS

CAS RN 12035-51-7

IUPAC Name: nickel disulfane
Synonyms: nickel disulfane
Molecular Weight (g/mol): 126.85
Molecular Formula: H4NiS2
InChi Key: CBJSXTNBQQMXNY-UHFFFAOYSA-N
SMILES: S.S.[Ni]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results