accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

KOO

CAS RN 12030-88-5

IUPAC Name: potassium dioxygen
Synonyms: potassium superoxide
Molecular Weight (g/mol): 71.1
Molecular Formula: KO2
InChi Key: CLSKHAYBTFRDOV-UHFFFAOYSA-N
SMILES: [K+].O=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)