accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OOHOOSiWWWOOOOOOWOOOWOOOWHOOOWOOOOOOOOWWOOOWOOHOOWOOOWOOHO

CAS RN 12027-43-9

IUPAC Name: tetrakis(hexaoxocyclotritungstoxane) silicic acid hydrate
Synonyms: silicic acid tetrakis(tungsten oxide) hydrate
Molecular Weight (g/mol): 2896.17
Molecular Formula: H6O41SiW12
InChi Key: ACTPFSYIGLFGLY-UHFFFAOYSA-N
SMILES: O.O[Si](O)(O)O.O=[W]1(=O)O[W](=O)(=O)O[W](=O)(=O)O1.O=[W]1(=O)O[W](=O)(=O)O[W](=O)(=O)O1.O=[W]1(=O)O[W](=O)(=O)O[W](=O)(=O)O1.O=[W]1(=O)O[W](=O)(=O)O[W](=O)(=O)O1
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results